Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAC1, Guinea pig, Polyclonal Antibody, Abnova™

Catalog No. 89126618 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-126-618 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-126-618 Supplier Abnova Corporation Supplier No. PAB24942
Add to Cart
Add to Cart

Guinea pig polyclonal antibody raised against synthetic peptide of Tac1.

Sequence: CRPKPQQFFGLM

Specifications

Antigen TAC1
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Guinea pig polyclonal antibody raised against synthetic peptide of Tac1.
Dilution Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user.
Formulation In antiserum (0.05% sodium azide)
Gene Tac1
Gene Alias PPTA3/Ppt5fl/RATPPTA3/TAC
Gene Symbols Tac1
Host Species Guinea Pig
Immunogen A synthetic peptide corresponding to amino acids 1-11 of rat Tac1.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24806
Target Species Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.