Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guinea Pig SerumRabbit anti-Guinea Pig, Polyclonal, Rockland™
SDP

Catalog No. 501990223
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0223 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-0223 Supplier Rockland Immunochemicals Supplier No. 1064101
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-rabbit serum and multiple precipitin arcs against Guinea Pig Serum.

Guinea Pig Serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health. Guinea Pig serum antibody is suitable for use in veterinary research and biotechnology applications and is ideal for investigators in Cancer and Cell Biology.

Specifications

Antigen Guinea Pig Serum
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 32 mg/mL
Conjugate Unconjugated
Formulation 0.01M sodium phosphate with 0.15M NaCl and no preservative; pH 7.2
Host Species Rabbit
Immunogen Guinea Pig serum proteins
Quantity 2 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Guinea Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.