Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Guinea Pig TNF-alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $655.00
Specifications
Antigen | TNF-alpha |
---|---|
Clone | 569826 |
Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | ELISA |
Classification | Monoclonal |
Description
TNF-alpha Monoclonal specifically detects TNF-alpha in Guinea Pig samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
TNF-alpha | |
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
TNF | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
569826 | |
ELISA | |
Unconjugated | |
Mouse | |
Guinea Pig | |
P51435 | |
7124 | |
E. coli-derived recombinant guinea pig TNF-alpha Leu79-Leu234 Accession # P51435 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects guinea pig TNF-alpha in ELISAs. In sandwich immunoassays, no cross-reactivity with human, mouse, rat, porcine, canine, feline, equine, bovine, or cotton rat TNF-alpha is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title