Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GULP1/CED-6 Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NB1001042
Description
GULP1/CED-6 Polyclonal specifically detects GULP1/CED-6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
GULP1/CED-6 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
CED6GULPCED-6, Cell death protein 6 homolog, engulfment adapter protein, FLJ31156, GULP, engulfment adaptor PTB domain containing 1, Protein GULP, PTB domain adapter protein CED-6, PTB domain adaptor protein CED-6, PTB domain-containing engulfment adapter protein 1 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Human. Expected from sequence similarity: Mouse, Rat and Dog. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
0.5 mg/mL | |
Western Blot 0.2 - 0.6 ug/ml, Immunohistochemistry 5 - 10 ug/ml, Immunohistochemistry-Paraffin 5 - 10 ug/ml, Peptide ELISA Detection limit 1:64000 | |
Q9UBP9 | |
GULP1 | |
Peptide with sequence EGTVFCLDPLDSRC corresponding to C-Terminus according to NP_057399.1. | |
0.1 mg | |
Apoptosis | |
51454 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction