Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GULP1/CED-6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | GULP1/CED-6 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
GULP1/CED-6 Polyclonal specifically detects GULP1/CED-6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
GULP1/CED-6 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
CED6GULPCED-6, Cell death protein 6 homolog, engulfment adapter protein, FLJ31156, GULP, engulfment adaptor PTB domain containing 1, Protein GULP, PTB domain adapter protein CED-6, PTB domain adaptor protein CED-6, PTB domain-containing engulfment adapter protein 1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-130 of human GULP1/CED-6 (NP_057399.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis | |
PBS with 50% glycerol, pH7.3. | |
51454 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title