Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GULP1 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518051
Description
This antibody is predicted to react with canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
The prompt clearance of cells undergoing apoptosis is critical during embryonic development, normal tissue turnover, inflammation, and autoimmunity. CED6 is an evolutionarily conserved adaptor protein required for efficient engulfment of apoptotic cells by phagocytes.
Specifications
GULP1 | |
Polyclonal | |
Unconjugated | |
GULP1 | |
3110030A04Rik; 5730529O06Rik; CED 6; Ced6; CED-6; CED-6 protein; Cell death protein 6 homolog; engulfment adapter protein; FLJ31156; GULP; GULP, engulfment adaptor PTB domain containing 1; GULP1; Gulp-2; OTTHUMP00000205908; OTTHUMP00000205909; OTTHUMP00000205911; Protein GULP; PTB domain adapter protein CED-6; PTB domain adaptor protein CED-6; PTB domain-containing engulfment adapter protein 1 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
51454 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9UBP9 | |
GULP1 | |
Synthetic peptide sequence (EGTVFCLDPLDSRC) corresponding to the C-terminus amino acids of GULP1 (aa 291-304). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction