Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gum Arabic, Spray-Dried Powder, FCC, Spectrum™ Chemical
SDP

Catalog No. p-7157527 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-089 500 g Poly Bottle
18-603-090 2.5 kg Poly Pail
18-603-091 12 kg Poly Pail
18-603-092 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-603-089 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. GU113500GM
Add to Cart
Add to Cart

GU113, 9000-01-5

Spectrum™ Chemical Gum Arabic, Spray-Dried Powder, FCC is used in the food industry as an emulsifier, a confectionary coating and as a soluble dietary fiber. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9000-01-5

Specifications

CAS Max % 100%
Ash 5%
Quantity 500 g
Grade FCC
Insoluble Matter 1%
Loss on Drying 15%
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.