Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GUSB Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5122210
This item is not returnable.
View return policy
Description
GUSB Polyclonal Antibody for Western Blot, ICC/IF, IHC (F), IHC (P)
The GUSB gene encodes beta-glucuronidase (EC 3.2.1.31), a lysosomal hydrolase involved in the stepwise degradation of glucuronic acid-containing glycosaminoglycans. It is a tetrameric glycoprotein composed of identical subunits. The GUSB gene is mutated in mucopolysaccharidosis type VII (MPS7; MIM 253220).
Specifications
GUSB | |
Polyclonal | |
Unconjugated | |
GUSB | |
Ac2-223; adipose storage deficiency; AI747421; asd; beta glucuronidase; beta-D-glucuronidase; beta-G1; Beta-glucuronidase; BG; g; glucuronidase beta; glucuronidase, beta; Gur; Gus; Gusb; Gus-r; Gus-s; Gus-t; Gus-u; Gut; lysosomal enzyme; MPS7; si:ch211-160e1.7; si:ct573103.7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2990 | |
-20°C | |
Liquid |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P08236 | |
GUSB | |
Recombinant protein Glucuronidase Beta. The antigen corresponds to amino acid range 451-649 of the target protein. | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction