Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GUSB Polyclonal Antibody

GUSB Polyclonal Antibody
Supplier: Thermo Scientific PA5106619
Description
GUSB Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| GUSB | |
| Polyclonal | |
| Unconjugated | |
| GUSB | |
| Ac2-223; adipose storage deficiency; AI747421; asd; beta glucuronidase; beta-D-glucuronidase; beta-G1; Beta-glucuronidase; BG; g; glucuronidase beta; glucuronidase, beta; Gur; Gus; Gusb; Gus-r; Gus-s; Gus-t; Gus-u; Gut; lysosomal enzyme; MPS7; si:ch211-160e1.7; si:ct573103.7 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 110006, 24434, 2990 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P06760, P08236, P12265 | |
| GUSB | |
| A synthesized peptide derived from human GUSB(Accession P08236), corresponding to amino acid residues I326-L376. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction