Learn More
3M™ GVP-Series Belt-Mounted PAPRs Compatible Breathing Tubes
Shop All 3M Products
Click to view available options
For Use With (Equipment):
GVP system with H series hood
Description
- For use with H Series Hoods, or GVP Systems that have 3M™ Full Facepieces 7800S and 6000 DIN Series.
- 3M Catalog# GVP-122
Specifications
Specifications
| Material | Vinyl |
| Product Type | Breathing Tube |
| For Use With (Application) | Assembly, machine operation |
| For Use With (Equipment) | GVP system with H series hood |
| Includes | Breathing tube, metal thumbscrew clamp and squeeze clamp |
| Model | GVP-122 |