Learn More
Specifications
Specifications
| Autoclavable | Autoclavable |
| Color | Transparent |
| Filter Type | GF/CA |
| Pore Size | 0.22 μm |
| Sterility | Non-sterile |
| For Use With (Application) | Analytical Sample Prep, HPLC; Dissolution testing; Content uniformity; Environmental samples; Composite assays; Analysis of Biological and Organic Fluids; Food Analysis |
| Temperature (English) Max. | 180°F |
| Temperature (Metric) Max | 82°C |
| Type | Syringe Filter |
| Diameter (Metric) | 33 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.