Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ GW 1929 hydrochloride
GREENER_CHOICESDP

Catalog No. 166450 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-645-0 50 mg
16-641-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 16-645-0 Supplier R&D Systems™ Supplier No. 1664/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective PPARγ agonist. Orally active

Highly selective orally active peroxisome proliferator-activated receptor (PPAR)γ agonist (pEC50 values are 8.05, < 4 and < 4 for human PPARγ, PPARα and PPARδ receptors respectively). Decreases glucose, fatty acid and triglyceride levels following oral administration in vivo.

Chemical Identifiers

CAS 1217466-21-1
Molecular Formula C30H30ClN3O4

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 532.03
Purity >98%
Molecular Weight (g/mol) 532.037
Chemical Name or Material GW 1929 hydrochloride
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.