Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GW 5074
GSA_VAGREENER_CHOICE

Catalog No. 138110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-811-0 10 mg
13-815-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 13-811-0 Supplier Tocris Bioscience™ Supplier No. 1381/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent, selective c-Raf1 kinase inhibitor

GW 5074 is a potent, selective and cell-permeable c-Raf1 kinase inhibitor (IC50 = 9 nM). GW 5074 displays ≥ 100-fold selectivity for raf kinase over CDK1, CDK2, c-src, ERK2, MEK, p38, Tie2, VEGFR2 and c-fm. GW 5074 binds mutant huntingtin (mHTT) and decreases levels of mHTT in cultured hippocampal neurons and fibroblasts from patients with Huntington's disease. Brain penetrant.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 220904-83-6
Molecular Formula C15H8Br2INO2

Specifications

Quantity 10 mg
Formula Weight 520.94
Purity >98%
Molecular Weight (g/mol) 520.95
Chemical Name or Material GW 5074
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.