Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GW 542573X
GREENER_CHOICE

Catalog No. 431110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-111-0 10 mg
43-115-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 43-111-0 Supplier Tocris Bioscience™ Supplier No. 4311/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective activator of KCa2.1 channels

Activator of small-conductance Ca2+-activated K+ channels (KCa2); selective for KCa2.1 (EC50 = 8.2 μM in HEK293 cells expressing hKCa2.1) with the profile hKCa2.1 > hKCa2.2 = hKCa2.3 > hKCa3.1 (IK).

Chemical Identifiers

CAS 660846-41-3
Molecular Formula C19H28N2O5

Specifications

Quantity 10 mg
Formula Weight 364.44
Purity >99%
Molecular Weight (g/mol) 364.442
Chemical Name or Material GW 542573X
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.