Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GW 6471
GREENER_CHOICE

Catalog No. 461810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-181-0 10 mg
46-185-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 46-181-0 Supplier Tocris Bioscience™ Supplier No. 4618/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

PPARα antagonist

PPARα antagonist (IC50 = 0.24 μM). Enhances the binding affinity of the PPARα ligand-binding domain to the co-repressor proteins SMRT and NCoR.

Chemical Identifiers

CAS 880635-03-0
Molecular Formula C35H36F3N3O4

Specifications

Quantity 10 mg
Formula Weight 619.67
Purity >98%
Molecular Weight (g/mol) 619.685
Chemical Name or Material GW 6471
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.