Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GW 788388
GREENER_CHOICE

Catalog No. 326410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-641-0 10 mg
32-645-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 32-641-0 Supplier Tocris Bioscience™ Supplier No. 3264/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective inhibitor of TGF-βRI

Potent inhibitor of transforming growth factor-β type I receptor (ALK5) (IC50 values are 18 and 93 nM for ALK5 binding and for TGF-β cellular assay respectively). Inhibits esophageal squamous cell carcinoma (ESCC)-induced neoangiogenesis. Orally active.

Chemical Identifiers

CAS 452342-67-5
Molecular Formula C25H23N5O2

Specifications

Quantity 10 mg
Formula Weight 425.48
Purity >98%
Molecular Weight (g/mol) 425.49
Chemical Name or Material GW 788388
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.