Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ GW 9508
GREENER_CHOICESDP

Catalog No. 264950 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-495-0 50 mg
26-491-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 26-495-0 Supplier R&D Systems™ Supplier No. 2649/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective FFA1 (GPR40) agonist

Potent and selective agonist for the free fatty acid receptor FFA1 (GPR40) (pEC50 values are 7.32, < 4.3 and < 4.3 for FFA1, FFA2 and FFA3 receptors respectively). Inactive against a range of other GPCRs, kinases, proteases, integrins and PPARs. Potentiates glucose-stimulated insulin secretion in MIN6 cells (pEC50 = 6.14).

Chemical Identifiers

CAS 885101-89-3
Molecular Formula C22H21NO3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 347.41
Purity >99%
Molecular Weight (g/mol) 347.41
Chemical Name or Material GW 9508
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.