Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GW 9662
GREENER_CHOICESDP

Catalog No. 150810 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-081-0 10 mg
15-085-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 15-081-0 Supplier Tocris Bioscience™ Supplier No. 1508/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective covalent PPARγ antagonist

GW 9662 is a selective irreversible PPARγ antagonist (IC50 values are 3.3, 32 and 2000 nM for PPARγ, PPARα and PPARδ respectively). Blocks the inhibition of osteoclast formation induced by IL-4 in the low micromolar range (1-2 μM), therefore is more potent than BADGE. Anticancer, inhibits growth of human mammary tumor cell lines.

Chemical Identifiers

CAS 22978-25-2
Molecular Formula C13H9ClN2O3

Specifications

Quantity 10 mg
Formula Weight 276.68
Purity >98%
Molecular Weight (g/mol) 276.676
Chemical Name or Material GW 9662
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.