Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GW182 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP128751C
Description
GW182 Polyclonal antibody specifically detects GW182 in Human, Mouse samples. It is validated for Western Blot, ImmunoblotSpecifications
GW182 | |
Polyclonal | |
Western Blot, Immunoblotting | |
CAG repeat protein 26, CAGH26FLJ22043, DKFZp666E117, EMSY interactor protein, Glycine-tryptophan protein of 182 kDa, GW182 autoantigen, GW182GW1, KIAA1460EDIE, MGC75384, Protein GW1, TNRC6, trinucleotide repeat containing 6, trinucleotide repeat containing 6A, trinucleotide repeat-containing gene 6A protein | |
The immunogen recognized by this antibody maps to a region between residue 575 and 625 of human GW182/glycine-tryptophan protein of 182 kDa (NP_055309.2). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunoblot | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
27327 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction