Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GW182 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP157134
Description
GW182 Polyclonal specifically detects GW182 in Human, Rat samples. It is validated for Western Blot.Specifications
GW182 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
CAG repeat protein 26, CAGH26FLJ22043, DKFZp666E117, EMSY interactor protein, Glycine-tryptophan protein of 182 kDa, GW182 autoantigen, GW182GW1, KIAA1460EDIE, MGC75384, Protein GW1, TNRC6, trinucleotide repeat containing 6, trinucleotide repeat containing 6A, trinucleotide repeat-containing gene 6A protein | |
Rabbit | |
Affinity purified | |
RUO | |
27327 | |
Centrifuge at 12,000 x g for 20 seconds. Reconstitute with 50 ul distilled water to a final concentration of 1.0 mg/ml in PBS buffer. Vortex followed by centrifuge again to pellet the solution. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q8NDV7 | |
TNRC6A | |
Synthetic peptides corresponding to GW182 (trinucleotide repeat containing 6A) The peptide sequence was selected from the N terminal of GW182. Peptide sequence RELEAKATKDVERNLSRDLVQEEEQLMEEKKKKKDDKKKKEAAQKKATEQ. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Equine: 100%; Mouse: 100%; Rat: 100%; Canine: 92%; Chicken: 85%. | |
Human, Rat, Canine, Equine, Equine | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction