Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GXYLT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119811
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119811 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119811 Supplier Invitrogen™ Supplier No. PA5119811
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: U937 cell lysate, HL-60 cell lysate, HUVEC cell lysate, human fetal skeletal muscle tissue. Subcellular Location: Membrane.

Glycosyltransferase which elongates the O-linked glucose attached to EGF-like repeats in the extracellular domain of Notch proteins by catalyzing the addition of xylose.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GXYLT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene GXYLT1
Gene Accession No. Q4G148
Gene Alias 242n2; Glt8d3; glucoside xylosyltransferase 1; glycosyltransferase 8 domain containing 3; glycosyltransferase 8 domain-containing protein 3; Glycosyltransferase 8 domain-containing protein 3-like protein; Gm1228; Gm87; GXYLT1; S33-D
Gene Symbols GXYLT1
Host Species Rabbit
Immunogen Synthetic peptide within human gxylt1 aa 400-440.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283464
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.