Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GYG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556489
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556489 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556489 Supplier Invitrogen™ Supplier No. PA556489
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FLGRVKPWNY TYDPKTKSVK SEAHDPNMTH PEFLILWWNI FTTNVLPLLQ QFGLVKDTCS YVNVEDVSGA ISHLSLGEIP AMAQPFVSSE ERKERWEQGQ ADYMGADSFD NIKRKLDTYL Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 75%.

This gene encodes a member of the glycogenin family. Glycogenin is a glycosyltransferase that catalyzes the formation of a short glucose polymer from uridine diphosphate glucose in an autoglucosylation reaction. This reaction is followed by elongation and branching of the polymer, catalyzed by glycogen synthase and branching enzyme, to form glycogen. This gene is expressed in muscle and other tissues. Mutations in this gene result in glycogen storage disease XV. This gene has pseudogenes on chromosomes 1, 8 and 13 respectively. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GYG1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GYG1
Gene Accession No. P46976
Gene Alias AU017667; glycogenin; glycogenin 1; glycogenin glucosyltransferase; glycogenin-1; Glycogenin-1-like protein; GN1; GN-1; GSD15; Gyg; GYG protein; Gyg1
Gene Symbols GYG1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GYG1. Recombinant protein control fragment (Product #RP-102158).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2992
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.