Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GYG1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116508
Description
Positive Samples: MCF7, SW480, Mouse skeletal muscle, Mouse kidney, Mouse liver, Rat heart Immunogen sequence: EAHDPNMTHP EFLILWWNIF TTNVLPLLQQ FGLVKDTCSY VNVEDVSGAI SHLSLGEIPA MAQPFVSSEE RKERWEQGQA DYMGADSFDN IKRKLDTYLQ.
This gene encodes a member of the glycogenin family. Glycogenin is a glycosyltransferase that catalyzes the formation of a short glucose polymer from uridine diphosphate glucose in an autoglucosylation reaction. This reaction is followed by elongation and branching of the polymer, catalyzed by glycogen synthase and branching enzyme, to form glycogen. This gene is expressed in muscle and other tissues. Mutations in this gene result in glycogen storage disease XV. This gene has pseudogenes on chromosomes 1, 8 and 13 respectively. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
GYG1 | |
Polyclonal | |
Unconjugated | |
GYG1 | |
AU017667; glycogenin; glycogenin 1; glycogenin glucosyltransferase; glycogenin-1; Glycogenin-1-like protein; GN1; GN-1; GSD15; Gyg; GYG protein; Gyg1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
27357, 2992, 81675 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.80 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O08730, P46976, Q9R062 | |
Gyg, GYG1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 234-333 of human GYG1 (NP_0011716491). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction