Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GYG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552156
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552156 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552156 Supplier Invitrogen™ Supplier No. PA552156
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PAFKQFGSSA KVVHFLGSMK PWNYKYNPQS GSVLEQGSAS SSQHQAAFLH LWWTVYQNNV LPLYKSVQAG EARASPGHTL CHSDVGGPCA DSASGVGEPC ENSTPSAGVP CANSPLGSNQ PAQGLPEPTQ IVDETLSL Highest antigen sequence identity to the following orthologs: Mouse - 26%, Rat - 33%.

GYG2 belongs to the glycogenin family. Glycogenin is a self-glycosylating protein involved in the initiation phase of glycogen biosynthesis. Multiple alternatively spliced transcript variants encoding different isoforms have been identified, including the predominant band around 66kDa and a light band around 45kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GYG2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GYG2
Gene Accession No. O15488
Gene Alias glycogenin 2; glycogenin glucosyltransferase; glycogenin-2; GN2; GN-2; GYG2
Gene Symbols GYG2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GYG2. Recombinant protein control fragment (Product #RP-92165).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8908
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.