Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GYKI 52466 dihydrochloride
GREENER_CHOICESDP

Catalog No. 145450 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-545-0 50 mg
14-541-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-545-0 Supplier Tocris Bioscience™ Supplier No. 1454/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective non-competitive AMPA antagonist

GYKI 52466 dihydrochloride is a selective noncompetitive AMPA receptor antagonist (IC50 values are 10-20, ∼ 450 and >50 μM for AMPA-, kainate- and NMDA-induced responses respectively). Skeletal muscle relaxant and orally-active anticonvulsant. Has anti-proliferative effects in transformed cells.

Chemical Identifiers

CAS 2319722-40-0
Molecular Formula C17H15N3O2

Specifications

Quantity 50 mg
Formula Weight 366.24
Purity >99%
Molecular Weight (g/mol) 293.326
Chemical Name or Material GYKI 52466 dihydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.