Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GYKI 53655 hydrochloride
GREENER_CHOICE

Catalog No. 255510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-551-0 10 mg
25-555-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 25-551-0 Supplier Tocris Bioscience™ Supplier No. 2555/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Non-competitive non-NMDA iGluR antagonist

Non-competitive AMPA and kainate receptor antagonist. Analog of GYKI 52466 (Cat. No. 1454). Prolongs the survival time after MgCl2- induced global cerebral ischemia. Exhibits anticonvulsant activity. Also blocks GluK3 homomeric receptors (IC50 = 63 μM) and GluK2b(R)/GluK3 heteroreceptors (IC50 = 32 μM) at high concentrations.

Chemical Identifiers

CAS 143692-48-2
Molecular Formula C19H21ClN4O3

Specifications

Quantity 10 mg
Formula Weight 388.85
Purity >99%
Molecular Weight (g/mol) 388.852
Chemical Name or Material GYKI 53655 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.