Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GYPA/GYPB Mouse anti-Human, Clone: HIR2, Abnova™

Catalog No. 89934221 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-221 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-221 Supplier Abnova Corporation Supplier No. MAB12957
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against human GYPA/GYPB.

Glycophorins A (GYPA) and B (GYPB) are major sialoglycoproteins of the human erythrocyte membrane which bear the antigenic determinants for the MN and Ss blood groups. GYPB gene consists of 5 exons and has 97% sequence homology with GYPA from the 5' UTR to the coding sequence encoding the first 45 amino acids. In addition to the M or N and S or s antigens, that commonly occur in all populations, about 40 related variant phenotypes have been identified. These variants include all the variants of the Miltenberger complex and several isoforms of Sta; also, Dantu, Sat, He, Mg, and deletion variants Ena, S-s-U- and Mk. Most of the variants are the result of gene recombinations between GYPA and GYPB. [provided by RefSeq]

Specifications

Antigen GYPA/GYPB
Applications Flow Cytometry, Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone HIR2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human GYPA/GYPB.
Dilution Flow Cytometry Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.09% sodium azide).
Gene Alias CD235a/GPA/GPErik/GPSAT/GpMiIII/HGpMiIII/HGpMiV/HGpMiX/HGpMiXI/HGpSta(C)/MN/MNS
Gene Symbols GYPA
Host Species Mouse
Immunogen Human peripheral blood mononuclear cell (PBMC).
Purification Method Protein G purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2993/2994
Target Species Human
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.