Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GYPA Mouse anti-Human, Clone: JC159, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB14809
Description
Mouse monoclonal antibody raised against native human GYPA.
- High specificity
- High reactivity
Specifications
GYPA | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.25-0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
P02724 | |
GYPA | |
Formalin fixed membrane from a Hairy cell leukemia. | |
100 μg | |
Primary | |
Human | |
Liquid |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
JC159 | |
Mouse monoclonal antibody raised against native human GYPA. | |
In 10 mM PBS. | |
CD235a/GPA/GPErik/GPSAT/GpMiIII/HGpMiIII/HGpMiV/HGpMiX/HGpMiXI/HGpSta(C)/MN/MNS | |
Mouse | |
Protein A/G purification | |
RUO | |
2993 | |
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction