Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GYPA Mouse anti-Human, Clone: SPM183, Abnova™

Catalog No. 89934217 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-217 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-217 Supplier Abnova Corporation Supplier No. MAB13035
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native human GYPA.

Glycophorins A (GYPA) and B (GYPB) are major sialoglycoproteins of the human erythrocyte membrane which bear the antigenic determinants for the MN and Ss blood groups. In addition to the M or N and S or s antigens that commonly occur in all populations, about 40 related variant phenotypes have been identified. These variants include all the variants of the Miltenberger complex and several isoforms of Sta, as well as Dantu, Sat, He, Mg, and deletion variants Ena, S-s-U- and Mk. Most of the variants are the result of gene recombinations between GYPA and GYPB. [provided by RefSeq]

Specifications

Antigen GYPA
Applications Agglutination, Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone SPM183
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native human GYPA.
Dilution Agglutination Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS.
Gene Alias CD235a/GPA/GPErik/GPSAT/GpMiIII/HGpMiIII/HGpMiV/HGpMiX/HGpMiXI/HGpSta(C)/MN/MNS
Gene Symbols GYPA
Host Species Mouse
Immunogen Human erythrocytes treated with neuraminidase.
Purification Method Polyethylene Glycol (PEG) precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2993
Target Species Human
Content And Storage Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.