Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GYPB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571844
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571844 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571844 Supplier Invitrogen™ Supplier No. PA571844
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GYPB Polyclonal Antibody for Western Blot, IHC (P)

This protein is a minor sialoglycoprotein in erythrocyte membranes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GYPB
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene GYPB
Gene Accession No. P06028
Gene Alias CD235b; glycophorin B (MNS blood group); glycophorin B/glycophorin A fusion protein; glycophorin Hop; glycophorin-B; GPB; GYP; GYPB; GYPB/GYPA fusion; hybrid glycophorin Kip; MNS; PAS-3; Sialoglycoprotein delta; SS; Ss blood group; SS-active sialoglycoprotein
Gene Symbols GYPB
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 22-51 amino acids from the Central region of human GYPB.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2994
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.