Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GYPB Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980644
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0644 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0644 Supplier Bioss Supplier No. BS10041R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This protein is a minor sialoglycoprotein in erythrocyte membranes.

Specifications

Antigen GYPB
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GYPB
Gene Accession No. P06028
Gene Alias CD235b; glycophorin B (MNS blood group); glycophorin B/glycophorin A fusion protein; glycophorin Hop; Glycophorin-B; GPB; GYP; GYPB; GYPB/GYPA fusion; hybrid glycophorin Kip; MNS; PAS-3; Sialoglycoprotein delta; SS; Ss blood group; SS-active sialoglycoprotein
Gene Symbols GYPB
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Glycophorin B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2994
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.