Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GYPB Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
| Antigen | GYPB |
|---|---|
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
GYPB Polyclonal specifically detects GYPB in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| GYPB | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 2994 | |
| IgG | |
| Affinity purified |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| CD235b antigen, glycophorin B (includes Ss blood group), glycophorin B (MNS blood group), glycophorin B (Ss blood group), glycophorin HeP2, glycophorin MiVI, glycophorin-B, GPB.NY, GPBHGpMiVI, GYPHe.NY, MNS, PAS-3, Sialoglycoprotein delta, Ss blood group, SS-active sialoglycoprotein, SSCD235b | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-91 of human GYPB (NP_002091.3). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title