Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GYPC Monoclonal Antibody (Ret40f)
GREENER_CHOICE

Catalog No. PIMA127131
Change view
Click to view available options
Quantity:
250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA127131 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA127131 Supplier Invitrogen™ Supplier No. MA127131
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control of tonsil suggested.

Glycophorin C (GYPC) is an integral membrane glycoprotein. It is a minor species carried by human erythrocytes, but plays an important role in regulating the mechanical stability of red cells. A number of glycophorin C mutations have been described. The Gerbich and Yus phenotypes are due to deletion of exon 3 and 2, respectively. The Webb and Duch antigens, also known as glycophorin D, result from single point mutations of the glycophorin C gene. The glycophorin C protein has very little homology with glycophorins A and B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GYPC
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone Ret40f
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Tissue culture supernatant with 0.09% sodium azide
Gene GYPC
Gene Accession No. P04921
Gene Alias 0610037F22Rik; CD236; CD236R; GE; Gerbich antigen; GLPC; Glycoconnectin; glycophorin C; glycophorin C (Gerbich blood group); glycophorin-C; Glycophorin-D; glycoprotein beta; GPC; GPD; GPYD; GYPC; GYPD; MGC117309; MGC126191; MGC126192; PAS-2; PAS-2'; Sialoglycoprotein D
Gene Symbols GYPC
Host Species Mouse
Immunogen Red cell ghosts.
Quantity 250 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2995
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.