Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GYPC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102401
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102401 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102401 Supplier Invitrogen™ Supplier No. PA5102401
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Glycophorin C/GPC.

Glycophorin C (GYPC) is an integral membrane glycoprotein. It is a minor species carried by human erythrocytes, but plays an important role in regulating the mechanical stability of red cells. A number of glycophorin C mutations have been described. The Gerbich and Yus phenotypes are due to deletion of exon 3 and 2, respectively. The Webb and Duch antigens, also known as glycophorin D, result from single point mutations of the glycophorin C gene. The glycophorin C protein has very little homology with glycophorins A and B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GYPC
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GYPC
Gene Accession No. P04921, Q6XFR6, Q78HU7
Gene Alias 0610037F22Rik; CD236; CD236R; GE; Gerbich antigen; GLPC; Glycoconnectin; glycophorin C; glycophorin C (Gerbich blood group); glycophorin-C; Glycophorin-D; glycoprotein beta; GPC; GPD; GPYD; GYPC; GYPD; MGC117309; MGC126191; MGC126192; PAS-2; PAS-2'; Sialoglycoprotein D
Gene Symbols GYPC
Host Species Rabbit
Immunogen A synthesized peptide derived from human GYPC(Accession P04921), corresponding to amino acid residues V72-S122.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2995, 364837, 71683
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.