Learn More
Eisco™ Gypsum Mineral Specimen
SCI_ED
Catalog No. S26644 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S26644 10/Pk.
S24689 1 Ea.
S24690 12/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S26644 Supplier Eisco™ Supplier No. ESNG0016PK10
Add to Cart
Add to Cart
Gypsum Mineral Specimen ideal for use in study of cleavage and fracture as example of imperfect cleavage planes.
  • For use in learning about mineral hardness
  • Gypsum is soft enough to be scratched by fingernail
  • One of the standards on Mohs hardness scale (value of 2)
  • Similar appearance to other minerals therefor useful in identification exercises
  • Crystals grow in soft, wide sheets, but is also commonly found filling veins and fractures in other rocks
Specifications
Product Type Mineral
Quantity 10/Pk.
Specimen Size 2 to 3 cm
Weight (Metric) 0.2 kg
Specimen Type Gypsum