Learn More
Eisco™ Gypsum Mineral Specimen
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Description
- For use in learning about mineral hardness
- Gypsum is soft enough to be scratched by fingernail
- One of the standards on Mohs hardness scale (value of 2)
- Similar appearance to other minerals therefor useful in identification exercises
- Crystals grow in soft, wide sheets, but is also commonly found filling veins and fractures in other rocks
Specifications
Specifications
Product Type | Mineral, Gypsum |
For Use With (Application) | Study of cleavage and fracture as example of imperfect cleavage planes |
Packaging | Clear labeled and barcoded bag |
Quantity | 1 Ea. |
Specimen Size | 2 to 3 cm |