Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GYS1 Monoclonal Antibody (3A7)
GREENER_CHOICE

Catalog No. PIMA515802
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515802 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515802 Supplier Invitrogen™ Supplier No. MA515802
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15802 targets GYS1 in indirect ELISA, FACS and WB applications and shows reactivity with Human samples. The MA5-15802 immunogen is purified recombinant fragment of human GYS1 expressed in E. Coli. MA5-15802 detects GYS1 which has a predicted molecular weight of approximately 85kDa.

Glycogen Synthase 1 (GYS1) catalyzes the addition of glucose monomers to the growing glycogen molecule through the formation of alpha-1,4-glycoside linkages. Mutations in this gene are associated with muscle glycogen storage disease. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GYS1
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 3A7
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene Gys1
Gene Accession No. P13807
Gene Alias EC 2.4.1.11; glycogen [starch] synthase, muscle; glycogen synthase; glycogen synthase 1; glycogen synthase 1 (muscle); glycogen synthase 1, muscle; glycogen synthase 3, brain; GSY; GYS; GYS1; Gys3; MGS
Gene Symbols Gys1
Host Species Mouse
Immunogen Purified recombinant fragment of human GYS1 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2997
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.