Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GYS1 Monoclonal Antibody (J.18.5)
GREENER_CHOICE

Catalog No. PIMA515022
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515022 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515022 Supplier Invitrogen™ Supplier No. MA515022
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

GYS1 Monoclonal Antibody (J.18.5) detects endogenous levels of total muscle and liver glycogen synthase protein. It is not recommended to aliquot this antibody.

Glycogen is a polysaccharide of glucose and serves as an energy storage in mammalian muscle and liver. Glycogen synthase catalyzes the rate-limiting step of glycogen biosynthesis through the addition of glucose monomers to the growing glycogen molecule through the formation of alpha-1,4-glycoside linkages. Glycogen synthase has two major isoforms in mammals: muscle isoform (GYS1) and liver isoform (GYS2).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GYS1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone J.18.5
Concentration 24 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100 μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5
Gene GYS1
Gene Accession No. A2RRU1, P13807, P17625, P54840, Q8VCB3, Q9Z1E4
Gene Alias BC021322; glycogen [starch] synthase, liver; Glycogen [starch] synthase, muscle; glycogen synthase 1; glycogen synthase 1 (muscle); glycogen synthase 1, muscle; glycogen synthase 2; glycogen synthase 2 (liver); glycogen synthase 3, brain; glycogen synthase, liver; GLYSN; GSY; GYS; Gys1; GYS2; Gys3; LGS; MGS
Gene Symbols GYS1, GYS2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the sequence of human glycogen synthase.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14936, 232493, 25623, 2997, 2998, 690987
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.