Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GYS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529739
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529739 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529739 Supplier Invitrogen™ Supplier No. PA529739
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A431, PC-12. Predicted reactivity: Mouse (98%), Rat (98%), Zebrafish (84%), Pig (99%), Rabbit (98%), Rhesus Monkey (99%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Glycogen Synthase 1 (GYS1) catalyzes the addition of glucose monomers to the growing glycogen molecule through the formation of alpha-1,4-glycoside linkages. Mutations in this gene are associated with muscle glycogen storage disease. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GYS1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene Gys1
Gene Accession No. A2RRU1, P13807
Gene Alias EC 2.4.1.11; glycogen [starch] synthase, muscle; glycogen synthase; glycogen synthase 1; glycogen synthase 1 (muscle); glycogen synthase 1, muscle; glycogen synthase 3, brain; GSY; GYS; GYS1; Gys3; MGS
Gene Symbols Gys1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Glycogen synthase 1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2997, 690987
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.