Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GYS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588921
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588921 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588921 Supplier Invitrogen™ Supplier No. PA588921
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DLLDWRYLGR YYQHARHLTL SRAFPDKFHV ELTSPPTTEG FKYPRPSSVP PSPSGSQASS PQSSDVEDEV EDERYDEEEE AERDRLNIKS PFSLSHVPHG KKKLHGEYKN; Positive Samples: Mouse liver.

The protein encoded by this gene, liver glycogen synthase, catalyzes the rate-limiting step in the synthesis of glycogen - the transfer of a glucose molecule from UDP-glucose to a terminal branch of the glycogen molecule. Mutations in this gene cause glycogen storage disease type 0 (GSD-0) - a rare type of early childhood fasting hypoglycemia with decreased liver glycogen content.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GYS2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene GYS2
Gene Accession No. Q8VCB3
Gene Alias BC021322; cb765; Glycogen [starch] synthase, liver; glycogen liver; glycogen synthase; glycogen synthase 2; glycogen synthase 2 (liver); glycogen synthase, liver; GLYSN; GYS2; LGS; zgc:112057
Gene Symbols GYS2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 594-703 of human GYS2 (NP_0687762).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 232493
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.