Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GZF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113570
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113570 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113570 Supplier Invitrogen™ Supplier No. PA5113570
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LCDVTVSVEY QGVRKDFMAH KAVLAATSKF FKEVFLNEKS VDGTRTNVYL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 86%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 92%; Rat: 100%.

The GDNF-inducible zinc finger protein 1 (GZF1) is a sequence-specific transcriptional repressor with a BTB/POZ domain and ten zinc finger motifs whose expression is required for renal branching morphogenesis during kidney development. GZF1 binds to the 5'regulatory region of the homeodomain protein HOXA10, suggesting that GZF1 may play a role in morphogenesis other than kidney development. Recent experiments have indicated that GZF1 associates with nucleolin and this association is mediated by the first four zinc finger motifs of GZF1. It is thought that Nucleolin modulates the subcellular localization of GZF1 as well as its transcriptional repressor activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GZF1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GZF1
Gene Accession No. Q9H116
Gene Alias GDNF inducible zinc finger protein 1; GDNF-inducible zinc finger protein 1; GZF1; ZBTB23; Zinc finger and BTB domain-containing protein 23; zinc finger protein 336; ZNF336
Gene Symbols GZF1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human ZNF336 (aa 30-79).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64412
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.