Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GZMB/GZMH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115323
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115323 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115323 Supplier Invitrogen™ Supplier No. PA5115323
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Granzyme B/H.

GZMB/GZMH are members of the granzyme subfamily of proteins, part of the peptidase S1 family of serine proteases. The encoded preproprotein is secreted by natural killer (NK) cells and cytotoxic T lymphocytes (CTLs) and proteolytically processed to generate the active protease, which induces target cell apoptosis. This protein also processes cytokines and degrades extracellular matrix proteins, and these roles are implicated in chronic inflammation and wound healing. Expression of this gene may be elevated in human patients with cardiac fibrosis. [provided by RefSeq, Sep 2016].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GZMB/GZMH
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Gzmb
Gene Accession No. P04187, P10144, P18291, P20718
Gene Alias AI553453; C11; cathepsin G-like 1; Cathepsin G-like 2; cathepsin G-like 2, protein h-CCPX; CCP1; CCP-1/C11; CCPI; CCP-X; CGL1; CGL-1; CGL2; CGL-2; CSPB; CSP-B; CSP-C; CTLA1; CTLA-1; CTSGL1; CTSGL2; Cytotoxic cell protease 1; cytotoxic serine protease B; Cytotoxic serine protease C; Cytotoxic T-lymphocyte proteinase; cytotoxic T-lymphocyte proteinase 2; cytotoxic T-lymphocyte-associated serine esterase 1; cytotoxin serine protease-C; fragmentin; fragmentin 2; fragmentin-2; GLP I; GLP III; GLP-1; granzyme 2; granzyme B; granzyme B (granzyme 2, cytotoxic T-lymphocyte-associated serine esterase 1); granzyme B(G,H); Granzyme H; granzyme H (cathepsin G-like 2, protein h-CCPX); Granzyme-2; granzyme-like protein 1; granzyme-like protein I; GRB; GZB; GZMB; GZMH; GZMH/GZMB; HLP; human lymphocyte protein; Lymphocyte protease; Natural killer cell protease 1; RNKP-1; SECT; T-cell serine protease 1-3E
Gene Symbols Gzmb, GZMH
Host Species Rabbit
Immunogen A synthesized peptide derived from human GZMB(Accession P10144), corresponding to amino acid residues M1-G50.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14939, 171528, 2999, 3002
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.