Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GZMB Mouse anti-Human, Unconjugated, Clone: GZMB/2403, Abnova™

Catalog No. 89337708 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-708 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-708 Supplier Abnova Corporation Supplier No. MAB21215
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

GZMB Monoclonal antibody specifically detects GZMB in Human samples. It is validated for IHC-P.

Specifications

Antigen GZMB
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone GZMB/2403
Conjugate Unconjugated
Description granzyme B (granzyme 2, cytotoxic T-lymphocyte-associated serine esterase 1)
Gene GZMB
Gene Alias CCPI, CGL-1, CGL1, CSP-B, CSPB, CTLA1, CTSGL1, HLP, SECT
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 73-187 of human GZMB
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3002
Target Species Human
Content And Storage Store at 4°C
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.