Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ H 151
GREENER_CHOICESDP

Catalog No. 667550 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-755-0 50 mg
66-751-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 66-755-0 Supplier Tocris Bioscience™ Supplier No. 6675/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

STING antagonist

H 151 is a STING antagonist. Covalently binds STING at Cys91, blocking palmitoylation and preventing assembly of STING clusters. Decreases inflammatory cytokine production in human and mouse cells in vitro. Also blocks CMA (Cat. No. 6677) -induced increases in levels of inflammatory cytokines in mice, and alleviates systemic inflammation in a mouse model of autoinflammatory disease.

Specifications

Chemical Name or Material N-(4-Ethylphenyl)-N'-1H-indol-3-yl-urea
CAS 941987-60-6
Synonym ERIS, hMITA, MITA, mitochondrial mediator of IRF3 activation, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner, sting, transmembrane protein 173
Quantity 50 mg
Target STING Antagonists
Molecular Formula C17H17N3O
Formula Weight 279.34
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.