Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H-2 Class I Mouse, anti-Mouse, R718, Clone: M1/42, BD OptiBuild™

Catalog No. BDB751875 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751875 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751875 Supplier BD Biosciences Supplier No. 751875
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The M1/42 monoclonal antibody (mAb) specifically recognizes intact Histocompatibility-2 Region (H-2) class I antigens (Ags), also known as mouse Major Histocompatibility Complex class I (MHC-I) Ags. This mAb reportedly binds to one or more of the H-2 Ags (H-2K, H-2D, and likely H-2L) that are primarily expressed on most nucleated somatic cells of hematopoietic and nonhematopoietic origin. H-2 class I molecules are comprised of a ∼45-50 kDa type I transmembrane heavy chain glycoprotein that is noncovalently linked to ∼13 kDa β2-microglobulin (β2-m). The extracellular region of the polymorphic heavy chains is comprised of three globular domains (α1, α2, and α3), followed by a transmembrane region and a cytoplasmic tail. Sequence variations are manifest in regions of the α1 and α2 domains that line the peptide-binding cleft involved in Ag presentation. The M1/42 mAb reportedly binds to H-2 class I Ags from multiple mouse haplotypes including a, b, d, j, k, s, and u. It does not bind to separated H-2 class I heavy chains or β2-m. Cell surface CD8 molecules bind to invariant sites of the H-2 heavy chains and can provide coreceptor signaling for Ag-mediated activation through the T cell receptor. H-2 class I molecules that present self-Ags can lead to self-tolerance of maturing CD8+ T cells due to thymic selection. Alternatively, these molecules can present foreign peptide Ags to mature peripheral CD8+ T cells resulting in cell-mediated immune responses against foreign Ags. H-2 class I Ags also serve as ligands for activating and inhibitory receptors expressed by NK cells and some T cells. The M1/42 antibody is useful for analyzing H-2 class I Ag expression on cells or cell lines. Cells from different experimental systems, eg, stressed cells that have undergone infection or transformation, may express little or no H-2 class I Ag. In contrast, cells undergoing activation or responding to certain factors (eg, interferons) may express upregulated levels of these Ags.

TRUSTED_SUSTAINABILITY

Specifications

Antigen H-2 Class I
Applications Flow Cytometry
Classification Monoclonal
Clone M1/42
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.