Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H-2 Class I Rat anti-Mouse , PE, Clone: M1/42 (also known as M1/42.3.9.8), BD Biosciences™

Catalog No. BDB566776 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566776 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB566776 Supplier BD Biosciences Supplier No. 566776
Add to Cart
Add to Cart

Rat Monoclonal Antibody

H-2 Class I Monoclonal antibody specifically detects H-2 Class I in Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen H-2 Class I
Applications Flow Cytometry
Classification Monoclonal
Clone M1/42 (also known as M1/42.3.9.8)
Conjugate PE
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias H-2 Class I; H2 Class I; Mouse MHC Class I; Mouse MHC-I
Host Species Rat
Immunogen Mouse B10 Spleen Cells
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions, and unconjugated antibody and free PE were removed.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.