Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H-2 Class I Rat Anti-Mouse, RB613, Clone: M1/42 (also known as M1/42.3.9.8), BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 758316
Description
H-2 Class I Monoclonal antibody specifically detects H-2 Class I in Mouse (Tested in Development) samples. It is validated for Flow Cytometry
Specifications
| H-2 Class I | |
| Monoclonal | |
| 0.2 mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| 50 μg; | |
| Primary | |
| Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| M1/42 (also known as M1/42.3.9.8) | |
| RB613 | |
| H-2 Class I; H2 Class I; Mouse MHC Class I; Mouse MHC-I | |
| Mouse B10 Spleen Cells | |
| RUO | |
| 111364 | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction