Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H-2Db Antibody (27-11-13S), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP128190
Description
H-2Db Monoclonal specifically detects H-2Db in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunocytochemistry.Specifications
H-2Db | |
Monoclonal | |
0.5mg/mL | |
Flow Cytometry 1 μg/106 cells | |
H2-D1 | |
BDF1 mouse splenocytes | |
0.25 mg | |
Primary | |
An epitope on the alpha chain of H-2D | |
Store at 4C. Do not freeze. | |
IgG2a κ |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunocytochemistry | |
27-11-13S | |
FITC | |
H2Db, MHC class I | |
Mouse | |
Protein A or G purified | |
RUO | |
14964 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction