Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H-2Db Mouse, anti-Mouse, R718, Clone: KH95, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 751937
Description
The KH95 antibody specifically recognizes the H-2D[b] MHC class I alloantigen. Reactivity with other haplotypes (e.g., a, d, f, k, n, p, q, r, s, u, v) has not been observed.

Specifications
H-2Db | |
Monoclonal | |
R718 | |
50 μg | |
Primary | |
Refrigerate Upon Arrival |
Flow Cytometry | |
KH95 | |
Mouse | |
RUO | |
Mouse | |
Liquid |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction