Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ H-2Dd Monoclonal Antibody (34-5-8S), FITC
Mouse Monoclonal Antibody
Supplier: Invitrogen™ A15444

Description
H-2Dd Monoclonal Antibody for Flow
Involved in the presentation of foreign antigens to the immune system. [UniProt] HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
Specifications
H-2Dd | |
Monoclonal | |
0.5 mg/mL | |
PBS with <0.1% sodium azide; pH 7.4 | |
P01895, P01896, P01897, P01899, P01900, P14426, P14427 | |
H2-D1 | |
mouse H-2Dd from BDF1 mouse splenocytes. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
Flow Cytometry | |
34-5-8S | |
FITC | |
H2-D1 | |
H-2 class I histocompatibility antigen, D-B alpha chain; H-2D; H2-D; H-2D cell surface glycoprotein; H-2D(B); H2D1; H2-D1; H2-DC1-beta transplantation antigen protein; H2DD; H2L; H2LD; histocompatibility 2, D region locus 1; MHC class I antigen; MHC class I H2 antigen; MHC class I H-2Dp; MHC H2-D-q alpha-chain; Q5k protein | |
Mouse | |
purified | |
RUO | |
14964 | |
4°C, store in dark | |
Liquid |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction